Please login to your online account to display your discounted pricing

Fisherbrand™ Colored Label Tape, 0.5in. Wide, 14yds. Long

0.5 in. (13mm) wide rolls available in a variety of colors. Fisherbrand™ Colored Label Tape is useful for many labeling applications. It can be written on and will adhere to most surfaces.

$24.33 - $24.33

Specifications

Format Tape
Product Type Colored Label Tape
Quantity 6 Pack
For Use With (Application) Waterproof, Oil and Acid resistant
Material Paper
View More Specs
Products
Catalog Number Mfr. No. Color Price Quantity    

15901J

  Accessories
fisherbrand™
15901J
Copper Pack of 6 Each for $24.33
Case of 4 Pack for $97.32

15901K

  Accessories
fisherbrand™
15901K
Gray Pack of 6 Each for $24.33
Case of 4 Pack for $97.32

15901L

  Accessories
fisherbrand™
15901L
Lavender Pack of 6 Each for $24.33
Case of 4 Pack for $97.32

15901M

  Accessories
fisherbrand™
15901M
Lime Pack of 6 Each for $24.33
Case of 4 Pack for $97.32

15901Q

  Accessories
fisherbrand™
15901Q
Violet Pack of 6 Each for $24.33
Case of 4 Pack for $97.32

15901N

  Accessories
fisherbrand™
15901N
Rose Pack of 6 Each for $24.33
Case of 4 Pack for $97.32

15901B

  Accessories
fisherbrand™
15901B
Yellow Pack of 6 Each for $24.33
Case of 4 Pack for $97.32

15901A

  Accessories
fisherbrand™
15901A
White Pack of 6 Each for $24.33
Case of 4 Pack for $97.32

15901C

  Accessories
fisherbrand™
15901C
Green Pack of 6 Each for $24.33
Case of 4 Pack for $97.32

15901D

  Accessories
fisherbrand™
15901D
Pink Pack of 6 Each for $24.33
Case of 4 Pack for $97.32

15901E

  Accessories
fisherbrand™
15901E
Red Pack of 6 Each for $24.33
Case of 4 Pack for $97.32

15901F

  Accessories
fisherbrand™
15901F
Orange Pack of 6 Each for $24.33
Case of 4 Pack for $97.32

15901G

  Accessories
fisherbrand™
15901G
Blue Pack of 6 Each for $24.33
Case of 4 Pack for $97.32

15901H

  Accessories
fisherbrand™
15901H
Aqua Pack of 6 Each for $24.33
Case of 4 Pack for $97.32
Description & Specifications

Specifications

Format Tape
Product Type Colored Label Tape
Quantity 6 Pack
For Use With (Application) Waterproof, Oil and Acid resistant
Material Paper
Temperature Range (English) -10 to 250°F
Temperature Range (Metric) -23 to 121°C
Width (English) 0.5 in.

  • Strong, durable, and oil-, water-, and acid-resistant
  • Excellent for color coding
  • Withstand temperatures from -23° to +121°C (-10° to +250°F)
  • Can be written on with pen, pencil, or permanent marker
  • Stick to most surfaces including glass, plastic, and metal
  • Remove cleanly and easily
  • Tapes are supplied without dispenser

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.