Please login to your online account to display your discounted pricing

Fisherbrand™ Color Coders for Externally and Internally Threaded Cryogenic Storage Vials


For sample identification

Manufacturer: fisherbrand™  N377251

 View more versions of this product

Catalog No. 12-567-505

  • / Case of 500 Each
 

Description & Specifications

Specifications

For Use With Internally Threaded Cryogenic Storage Vials (12-567-500, -501, -502)
Material Polystyrene
Color Green
Sterile No