missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Color-Coded Biohazard Specimen Transport Bags
Color-coded bag lip for easy identification
Manufacturer: Fisherbrand™ RDQ607
Description
- Measure 6 × 9 in. (15 × 23cm)
- Printed with orange biohazard symbol, blue stat and storage instructions
- Document pouch keeps paperwork separate from specimens
Order Info
Packaged 100 per pack, 10 packs per case; assorted case contains 200 bags of each color
Specifications
Zip closure | |
9 in. | |
6 in. | |
Blue | |
PE (Polyethylene) |
9 x 6 in. (23 x 15cm) | |
Non-sterile | |
no | |
23cm | |
15cm |