missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Color-Coded Biohazard Specimen Transport Bags
Color-coded bag lip for easy identification
$125.76 - $215.79
Specifications
Closure Type | Zip Seal |
---|---|
Dimensions (L x W) | 9 x 6 in. (23 x 15cm) |
Length (English) | 9 in. |
Length (Metric) | 23 cm |
Width (English) | 6 in. |
Catalog Number | Mfr. No. | Color | Material | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Material | Price | Quantity | ||||
01-800-10
|
N/A | Blue | Polyethylene |
Case of 1000 Each for $125.76
|
|
||||
01-800-11
|
N/A | Green | Polyethylene |
Case of 1000 Each for $125.76
|
|
||||
01-800-12
|
N/A | Orange | Polyethylene |
Case of 1000 Each for $125.76
|
|
||||
01-800-13
|
N/A | Red | Polyethylene |
Case of 1000 Each for $125.76
|
|
||||
01-800-14
|
N/A | Yellow | Polyethylene |
Case of 1000 Each for $125.76
|
|
||||
01-800-15
|
N/A | Assorted | LDPE |
Case of 1000 Each for $215.79
|
|
||||
Description
- Measure 6 x 9 in. (15 x 23cm)
- Printed with orange biohazard symbol, blue stat and storage instructions
- Document pouch keeps paperwork separate from specimens
Order Info
Packaged 100 per pack, 10 packs per case; assorted case contains 200 bags of each color
Specifications
Zip Seal | |
9 in. | |
6 in. |
9 x 6 in. (23 x 15cm) | |
23 cm | |
15 cm |