missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Color-Coded Biohazard Specimen Transport Bags
Color-coded bag lip for easy identification
Manufacturer: Fisherbrand™ Q612
Description
- Measure 6 × 9 in. (15 × 23cm)
- Printed with orange biohazard symbol, blue stat and storage instructions
- Document pouch keeps paperwork separate from specimens
Order Info
Packaged 100 per pack, 10 packs per case; assorted case contains 200 bags of each color
Specifications
9 x 6 in. (23 x 15cm) | |
100 | |
1.5 lb. | |
Zip closure | |
Document Compartment | |
23cm | |
6 in. |
1000 | |
No | |
.68kg | |
Assorted | |
9 in. | |
LDPE (Low-density Polyethylene) | |
15cm |