missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Color-Coded Biohazard Specimen Transport Bags

Color-coded bag lip for easy identification

Supplier:  Fisherbrand™ Q609

 View more versions of this product

Catalog No. 01-800-12



Description

Description

  • Measure 6 × 9 in. (15 × 23cm)
  • Printed with orange biohazard symbol, blue stat and storage instructions
  • Document pouch keeps paperwork separate from specimens

Order Info

Packaged 100 per pack, 10 packs per case; assorted case contains 200 bags of each color

Specifications

Specifications

Not Autoclavable
Orange
no
9 in.
6 in.
Zip Closure
Polyethylene
9 x 6 in. (23 x 15cm)
23 cm
15 cm
SDS
Documents

Documents

Promotions

Promotions