missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Color-Coded Biohazard Specimen Transport Bags
Color-coded bag lip for easy identification
Description
- Measure 6 × 9 in. (15 × 23cm)
- Printed with orange biohazard symbol, blue stat and storage instructions
- Document pouch keeps paperwork separate from specimens
Order Info
Packaged 100 per pack, 10 packs per case; assorted case contains 200 bags of each color
Specifications
| Not Autoclavable | |
| Green | |
| Non-sterile | |
| 9 in. | |
| 6 in. |
| Zip Seal | |
| Polyethylene | |
| 9 x 6 in. (23 x 15cm) | |
| 23 cm | |
| 15 cm |