Fisherbrand™ Color-Coded Autoclavable Identification Tape
Instrument marking tape for easy identification and color coding items. Even sticks to PTFE.
Manufacturer:  Fisherbrand™ 1590130F
 View more versions of this product
Catalog No. 1590130F
Description
Description
Instrument marking tape for easy identification and color coding items. Even sticks to PTFE.
  • Vinyl self-stick tapes color-code autoclave items such as surgical instruments for easy identification
  • Attach securely and safely to any clean surface
  • Resist water, oil, and acids
  • Nontoxic
Order Info
In 16 ft.L x 0.37 in. W (5m x 9.6mm) rolls.
Specifications
Specifications
Yes
Resist Water, Oil and Acids
4.8m
Tape
Identification Tape
Blue, Blue
16 ft.
0.37 in.
Vinyl, Vinyl
Each
Documents