Please login to your online account to display your discounted pricing
Fisherbrand™ Clear Straight-Sided Jars with PTFE-Faced PE-Lined Cap
Clear multipurpose jars for maximum content visibility. Qorpak™ Clear Straight-Sided Jars with PTFE-Faced PE-Lined Cap are available in in a wide variety of sizes, from 1 to 32 oz.
$74.97 - $612.37
Specifications
Closure Liner Polyethylene
Closure Material Polypropylene
Closure Screw
Color Clear
Material Glass
View More Specs
Products
Catalog Number Mfr. No. Capacity Packaging Closure Size Price Quantity    
02-911-761
fisherbrand™
264026
16 oz., 473mL 12/Cs. 89-400 Case of 12 Each for $106.40
02-911-762
fisherbrand™
264036
32 oz., 1000mL 12/Cs. 89-400 Case of 12 Each for $130.57
02-911-763
fisherbrand™
264034
2 oz., 60mL 24/Cs. 53-400 Case of 24 Each for $88.43
02-911-765
fisherbrand™
264044
8 oz., 250mL 12/Cs. 70-400 Case of 12 Each for $74.97
02-911-764
fisherbrand™
264041
4 oz., 125mL 24/Cs. 58-400 Case of 24 Each for $97.87
02-911-964
fisherbrand™
264074
2 oz., 60mL Case of 180 Each for $612.37
Description & Specifications
Specifications
Closure Liner Polyethylene
Closure Material Polypropylene
Closure Screw
Color Clear
Material Glass
Mouth Wide
  • Designed without shoulders for maximum storage capacity
  • Wide mouth for easy filling
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.