Please login to your online account to display your discounted pricing
Fisherbrand™ Clear Straight-Sided Jars with Polyvinyl-Lined Cap
For easy viewing of contents. Fisherbrand™ Clear Straight-Sided Jars with Polyvinyl-Lined Cap are for multipurpose use.
$32.57 - $120.90
Specifications
Closure Liner Poly/Vinyl
Closure Material Polypropylene
Closure Screw
Color Clear
Material Glass
View More Specs
Products
Catalog Number Mfr. No. Capacity Closure Size Price Quantity    
02-911-774
fisherbrand™
264037*
32 oz., 1000mL 89-400 Case of 12 Each for $75.40
02-911-773
fisherbrand™
264035
2 oz., 60mL 53-400 Case of 24 Each for $54.67
02-911-772
fisherbrand™
264027
16 oz., 473mL 89-400 Case of 12 Each for $55.73
02-911-775
fisherbrand™
264042
4 oz., 125mL 58-400 Case of 24 Each for $64.70
02-911-776
fisherbrand™
264045
8 oz., 250mL 70-400 Case of 12 Each for $32.57
02-911-920
fisherbrand™
264032
1 oz., 30mL 43-400 Case of 48 Each for $120.90
Description & Specifications
Specifications
Closure Liner Poly/Vinyl
Closure Material Polypropylene
Closure Screw
Color Clear
Material Glass
Mouth Wide
  • Designed without shoulders for maximum storage capacity
  • Available in a wide variety of sizes, from 1 oz. to 32 oz.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.