Please login to your online account to display your discounted pricing
Fisherbrand™ Clear PET Packer Bottles with White Polypropylene PE Foam Cap
Ideal for oil analysis. Fisherbrand™ Clear PET Packer Bottles with White Polypropylene PE Foam Cap provides a good alcohol and solvent barrier.
$65.10 - $104.70
Specifications
Certifications/Compliance ISO 9001
Closure Material Polypropylene
Closure Type Screw
Color Clear
Material PET
View More Specs
Products
Catalog Number Mfr. No. Capacity Closure Liner Closure Size Neck Finish Price Quantity    
02-911-795
fisherbrand™
264001
4.2 oz., 125mL Foam Polyethylene 38-400 38-400 Case of 48 Each for $67.13
02-911-808
fisherbrand™
264010
8.45 oz., 250mL Foam Polyethylene 45-400 45-400 Case of 48 Each for $104.70
02-911-849
fisherbrand™
264021
2 oz., 60mL Foam Polyethylene 33-400 33-400 Case of 48 Each for $65.10
Description & Specifications
Specifications
Certifications/Compliance ISO 9001
Closure Material Polypropylene
Closure Type Screw
Color Clear
Material PET
  • Clear and transparent for easy sample identification
  • PET is stress and impact resistant
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.