missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Graduated Glass Bottle Beakers with White Polypropylene PE Foam/PTFE Cap

Environmentally sensitive bottles help eliminate waste. Fisherbrand™ Clear Graduated Clear Graduated Boston Round Bottles with White Polypropylene PE Foam/PTFE Lined Cap ensures product integrity for long term storage.

Manufacturer:  Fisherbrand™ ALP263487

 View more versions of this product

Catalog No. 02-911-461A


 

Includes: Polypropylene PE Foam/PTFE lined cap

Description

Description

Graduated Medium Rounds are excellent multipurpose containers for use with biological and pathological specimens, but can also be used for storing industrial laboratory chemicals and reagents. These clear (flint) glass bottles are designed with a slight shoulder for easy pouring and handling. The wide mouth facilitates easy mixing, handling, and transfer of contents. Graduated in both milliliters and ounces.

Clear Graduated Boston Round Bottles
  • Feature a narrow screw neck opening, curved shoulders, and a round body
  • Glass offers minimal chemical interference with contents
  • Clear (flint) glass is appropriate when visibility is important but protection from UV rays is not necessary
 Polypropylene PE Foam/PTFE Lined Caps
  • Offer good impact strength, thermal resistance and pliability
  • Autoclavable
  • Liners consist of 3 plys: a low density foam core tightly sandwiched between two layers of polyethylene (LDPE)
  • Liners are very resilient and have strong resistance to chemicals and moisture and help keep bottle or jar contents free of odor and taste interference
Specifications

Specifications

8 oz., 240mL
8 oz.
ClearþClear
GlassþGlass
Bottle, GraduatedþGraduated Medium Round Bottle
PE Foam/PTFE
58-400
Screw
240mL
Polypropylene
Yes
24/Cs.
White
Wide
Round
Documents