Please login to your online account to display your discounted pricing
Fisherbrand™ Clear Square Tablet Bottles with Black Phenolic PolyCone Cap
Provides maximum content visibility. Fisherbrand™ Clear Square Tablet Bottles with Black Phenolic PolyCone Cap is available in capacities of 1 and 2 oz. and screw cap size of 33mm-400.
$463.50 - $741.47
Specifications
Closure Liner Polyethylene
Closure Material Phenolic
Closure Type Screw
Closure Color Black
Closure Size 33-400
View More Specs
Products
Catalog Number Mfr. No. Diameter (Metric) Capacity (Metric) Price Quantity    
02-911-993
fisherbrand™
264072
Case of 288 Each for $741.47
02-912-308
fisherbrand™
264073
Case of 144 Each for $463.50
Description & Specifications
Specifications
Closure Liner Polyethylene
Closure Material Phenolic
Closure Type Screw
Closure Color Black
Closure Size 33-400
Color Clear
Material Glass
  • Feature a space saving design for bench tops with limited storing space
  • Wide mouth bottles are ideal for mixing, storing and sampling
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.