Fisherbrand™ Clear Glass Jugs, Rubber Cap Lining
Sample visibility and premium quality
$62.43 - $64.23
Specifications
Closure Type Screw,Screw
Closure Size 38-400
Color Clear
Material Glass
Mouth Narrow
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity Price Quantity  
Catalog Number Mfr. No. Capacity Price Quantity  
02-912-274 Fisherbrand™
263989
130 oz., 3844.56mL Case of 4 Each for $64.23
02-912-275 Fisherbrand™
263993
64 oz., 1892.71mL Case of 6 Each for $62.43
Specifications
Description
  • Type III soda-lime glass
  • Narrow mouth enables careful pouring and prevents spillage
  • 38mm-400 screw caps attached
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
Screw,Screw
Clear
Narrow
38-400
Glass