Fisherbrand™ Clear French Square Bottles without Cap
Ideal for mixing, storing and sampling. Fisherbrand™ Clear French Square Bottles without Cap are space-saving and easy to fill.
Manufacturer:  Fisherbrand™ 263971
 View more versions of this product
Catalog No. 02-911-887
Description
Description
Clear (Flint) Glass French Square Bottles are space-saving and easy to fill. The clear glass provides maximum content visibility and minimal chemical inference with bottle’s contents. The French Square bottle design allows the bottle to hold the maximum volume of contents while taking up minimum shelf and storage space. The wide mouth opening is ideal for mixing, storing and sampling.
  • Features wide mouth opening
  • Clear glass provides maximum content visibility and minimal chemical inference with bottle contents
  • Allows the bottle to hold the maximum volume of contents while taking up minimum shelf and storage space
Specifications
Specifications
4 oz., 125mL
4 oz.
GlassþGlass
Bottle, French SquareþFrench Square Bottle
Wide
French Square
120mL
ClearþClear
120/Cs.
33-400
33-400
Screw
Documents
Accessories