Fisherbrand™ Clear French Square Bottles with White Polypropylene PE Foam/PTFE Lined Cap
Space-saving and easy to fill. Fisherbrand™ Clear French Square Bottles with White Polypropylene PE Foam/PTFE Lined Caps features wide mouth opening ideal for mixing, storing and sampling.
$86.80 - $151.57
Specifications
Material Glass
Closure Material Polypropylene
Closure Type Screw
Color Clear
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
02-911-385 Fisherbrand™
263947
15mL Case of 48 Each for $115.67
02-911-387 Fisherbrand™
263950
60mL Case of 48 Each for $113.57
02-911-386 Fisherbrand™
263949
30mL Case of 48 Each for $146.60
02-911-388 Fisherbrand™
263951
120mL Case of 24 Each for $86.80
02-911-389 Fisherbrand™
263952
240mL Case of 24 Each for $110.23
02-911-390 Fisherbrand™
263948
480mL Case of 24 Each for $151.57
02-912-016 Fisherbrand™
263946
960mL Case of 12 Each for $87.37
Specifications
Description
Clear French Square Bottles
  • Clear glass provides maximum content visibility and minimal chemical inference with bottle’s contents
  • French Square design allows bottle to hold the maximum volume of contents while taking up minimum shelf and storage space
Polypropylene PE Foam/PTFE Lined Cap
  • Offer good impact strength and pliability
  • Can be remelted after they are formed
  • Foam backed PTFE liners resist attack from virtually all chemicals at room temperature and provides resilience for a tight seal with a low moisture transmission rate
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
Glass
Polypropylene
Screw
Clear