Fisherbrand™ Clear Boston Round Bottles with Black Phenolic Rubber Cap
For storing liquid samples. Fisherbrand™ Boston Round Bottles with Black Phenolic Rubber Cap are clear for easy viewing of contents.
Manufacturer:  Fisherbrand™ W014012
 View more versions of this product
Catalog No. 02-911-756
Specifications
Description
Bottle, Boston round; Fisherbrand; Clear; Narrow-mouth; With attached rubber-lined closures; Type III soda lime glass; Capacity: 1 oz. (30mL); 48/case; 20mm-400 screw cap size
  • Features a narrow screw neck opening, curved shoulders, and a round body
  • Glass offers minimal chemical interference with contents
Description & Specifications
Specifications
1 oz., 30mL
1 oz.
Rubber
Soda Lime Glass
Screw
30mL
Rubber
Clear
20-400