Please login to your online account to display your discounted pricing

Fisherbrand™ Clear Boston Round Bottles with Black Phenolic Rubber Cap


For storing liquid samples. Fisherbrand™ Boston Round Bottles with Black Phenolic Rubber Cap are clear for easy viewing of contents.

Manufacturer: fisherbrand™  263922

 View more versions of this product

Catalog No. 02-911-759

  • / Case of 24 Each
 

Description & Specifications

Specifications

Capacity 4 oz., 125mL
Capacity (Metric) 120mL
Capacity (English) 4 oz.
Closure Material Phenolic
Closure Liner Rubber
Color Clear
Material Glass
Quantity 24/Cs.
Type Bottle, Boston Round
Closure Color Black
Closure Size 22-400
Liner Material Rubber
Mouth Narrow
Neck Finish 22-400
Shape Boston Round
Closure Type Screw

  • Features a narrow screw neck opening, curved shoulders, and a round body
  • Glass offers minimal chemical interference with contents