Fisherbrand™ Clear Boston Round Bottles with PTFE-Faced PE-Lined Cap
For storing liquid samples. Fisherbrand™ Clear Boston Round Bottles with PTFE-Faced PE-Lined Cap are ideal for use when content integrity is important.
$34.73 - $71.60
Specifications
Closure Liner PTFE/Polyethylene
Closure Type Screw
Closure Size 22-400
Color Clear
Material Glass
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
02-911-753 Fisherbrand™
263875
480mL Case of 12 Each for $53.07
02-911-754 Fisherbrand™
263907
960mL Case of 12 Each for $71.23
02-911-787 Fisherbrand™
263896
60mL Case of 24 Each for $47.63
02-911-789 Fisherbrand™
263936
240mL Case of 12 Each for $34.73
02-911-788 Fisherbrand™
263923
120mL Case of 24 Each for $71.60
Specifications
Description
  • Features a narrow screw neck opening, curved shoulders, and a round body
  • Glass offers minimal chemical interference with contents
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
PTFE/Polyethylene
22-400
Glass
Screw
Clear