Please login to your online account to display your discounted pricing

Fisherbrand™ Clear Boston Round Bottles with White Polypropylene Pulp/Vinyl Cap


Ideal general purpose bottle. Fisherbrand™ Clear Boston Round Bottles with White Polypropylene Pulp/Vinyl Cap ensure integrity for long term storage.

Manufacturer: fisherbrand™  263924

 View more versions of this product

Catalog No. 02-911-730

  • / Case of 160 Each
 

Description & Specifications

Specifications

Product Type Boston Round Bottle
Neck Finish 22-400
Shape Boston Round
Mouth Narrow
Material Glass
Mouth Style Narrow
Capacity (Metric) 120mL
Closure Material Polypropylene
Closure Color White
Closure screw cap
Closure Type Screw
Liner Material Pulp/Vinyl
Closure Included Yes
Capacity (English) 4 oz.
Color Clear
Quantity 160/Cs.
Packaging Type 160/Cs.
Capacity 4 oz., 125mL
Closure Liner Poly/Vinyl
Closure Size 22-400

  • Features narrow screw neck opening, curved shoulders and round body