Please login to your online account to display your discounted pricing

Fisherbrand™ Clear Boston Rounds with Polyvinyl-Lined Caps

Economical, high-quality bottles for storing liquid samples

Manufacturer: fisherbrand™  W014024

 View all options for this product

Catalog No. 02-911-781


Description & Specifications


Capacity 8 oz., 250mL
Capacity (Metric) 240mL
Capacity (English) 8oz.
Closure Color White
Closure Screw
Closure Material Polypropylene
Closure Liner Poly/Vinyl
Closure Size 24-400
Closure Type Screw
Color Clear
Material Glass
Liner Material Pulp/Vinyl
Mouth Narrow
Neck Finish 24-400
Packaging Type 12/Cs.
Quantity 12/Cs.
Shape Boston Round
Product Type Boston Round Bottle

  • Type III soda-lime glass
  • Clear for easy viewing of contents
  • Narrow mouth