Please login to your online account to display your discounted pricing

Fisherbrand™ Clear Automatic Replacement Burets for 03-842 Series

For 03-842-1 and 03-842-2 Series

$270.20 - $283.87

Specifications

Color Clear
Product Type Automatic Replacement Buret
For Use With (Equipment) For 03-842-1 and 03-842-2 series
Includes Glass stopcocks
Material Glass
View More Specs
Products
Catalog Number Mfr. No. Capacity (Metric) Price Quantity    

03-842-3A

 
fisherbrand™
7038423A
10mL Each for $270.20

03-842-3B

 
fisherbrand™
7038423B
25mL Each for $283.87
Description & Specifications

Specifications

Color Clear
Product Type Automatic Replacement Buret
For Use With (Equipment) For 03-842-1 and 03-842-2 series
Includes Glass stopcocks
Material Glass

  • With standard taper glass stopcocks
  • Capacity: 10 and 25mL