Fisherbrand™ Class B Cylinders with Single English Scale, Graduated in Ounces
Calibrated “To Contain”
Supplier:  Fisherbrand™ 08563B
 View more versions of this product
Catalog No. 08-563B
Description
Description
  • Accuracy assured even at lowest graduations
  • Zero at bottom
Specifications
Specifications
Round
Graduated Cylinder
4 oz.
Round
Glass
Single
Autoclavable
+/- 0.3 drams
±0.3 mL
To Contain
White
B
118.2 mL
1.7 mL
4 oz.
1.7mL
SDS
Documents
Documents
Accessories