Please login to your online account to display your discounted pricing

Fisherbrand™ Class B Clear Glass Threaded Vials


General purpose vial avoids alteration in pH of sample

Manufacturer: fisherbrand™  KFS60910D2

 View more versions of this product

Catalog No. 03-339-22D

  • / Pack of 144 Each
  • / Case of 12 Pack
 

Description & Specifications

Specifications

Material (Liner) Rubber
Length (Metric) 60mm
Diameter (Metric) Outside 17mm
For Use With (Application) For general purpose
Material Borosilicate Glass
Autoclavable Yes
Classification B
Material Closure Phenolic
Capacity (English) 2 dr.
Type Sample Vial
Color Clear
Volume (Metric) 7.4mL
Closure Type Closed screw top
Volume (English) 2 dr.