missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Class B Clear Glass Threaded Vials

General purpose vial avoids alteration in pH of sample

Manufacturer:  Fisherbrand™ FS60910D6

 View more versions of this product

Catalog No. 03-339-22G



 

Specifications Description & Specifications

Specifications

Yes
B
Clear
Borosilicate Glass
85mm
Phenolic
Rubber
Closed screw top
23mm
6 dr.
Sample Vial
22.2mL
For general purpose
6 dr.