Please login to your online account to display your discounted pricing

Fisherbrand™ Class B Clear Glass Threaded Vials


General purpose vial avoids alteration in pH of sample

Manufacturer: fisherbrand™  KFS60910D12

 View more versions of this product

Catalog No. 03-339-22A

  • / Pack of 144 Each
  • / Case of 16 Pack
 

Description & Specifications

Specifications

Autoclavable Yes
Closure Type Screw cap
Classification B
Diameter (Metric) Outside 12mm
Color Clear
Capacity (English) .5 dr.
Material Borosilicate Glass
Type Sample Vial
Length (Metric) 35mm
Volume (Metric) 1.8mL
Material Closure Phenolic
For Use With (Application) For general purpose
Material (Liner) Rubber
Volume (English) 0.5 dr.