Please login to your online account to display your discounted pricing

Round Clear Glass Bottles with PTFE Lined Cap


Manufacturer: fisher scientific  02912315

 View more versions of this product

Catalog No. 02-912-315

  • / Case of 24 Each
 

Description & Specifications

Specifications

Capacity (Metric) 250mL
Capacity (English) 8 oz.
Closure Included Yes
Closure Color White
Closure screw cap
Closure Material Polyethylene
Closure Size 45-400
Color Clear
Material Glass
Liner Material PTFE
Mouth Style Wide
Quantity 12/PK.
Capacity 8 oz., 250mL