Fisherbrand™ Certified Cleaned Amber Boston Round Bottles
Ideal storage for solvents, chemicals and samples. Fisherbrand™ Certified Cleaned Amber Boston Round Bottles are manufactured from amber colored soda glass.
Manufacturer:  Fisherbrand™ 02911928
 View more versions of this product
Catalog No. 02-911-928
Description
Description
  • Narrow mouth
  • Cleaned per EPA guidelines for semi-volatiles, pesticides, PCBs and metals
  • With white polypropylene cap with PTFE faced foamed polyethylene liner
Specifications
Specifications
8.45 oz., 250mL
8.45 oz.
EPA guidelines
Polyethylene
Glass
White
Round
250mL
Certificate of analysis
Polyethylene
Amber
Bottle, Boston Round
24-400
Screw