Please login to your online account to display your discounted pricing

Fisherbrand™ Cell Spreaders


Simplify bacterial culture work and prevent agar surface tears.

Manufacturer: fisherbrand™  6401SEMD

 View more versions of this product

Catalog No. S66131

  • / Pack of 25 Each
  • / Case of 4 Pack
 

Description & Specifications

Specifications

Autoclavable Yes
Color Blue
For Use With (Application) Simplify bacterial culture work and prevent agar surface tears, easy to manipulate around curves and into corners of prepared plates
For Use With (Equipment) Prepared biplates
Material Polypropylene
Product Type Cell Spreader
Sterile Yes