missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Cell Scrapers
Easily harvest cells from tissue culture flasks
Manufacturer: Fisherbrand™ GDPC241
Description
Easily harvest cells from tissue culture flasks
Fisherbrand Cell Scrapers are appropriate for a variety of cell culture vessels. The uniquely designed handle and blade reduce cell damage during the harvesting process.
- Presterilized and individually wrapped in peel-open packaging
- Nonpyrogenic
- Polyethylene copolymer blade pivots to permit even contact with growth surface
- Ribbed polystyrene handle provides secure grip and added control
- Flexible joint between blade and handle eases entry into vessel
- Pivoting blade improves position control to adjust for difficult angles during cell harvesting
Specifications
For a variety of cell culture vessels | |
0.7 in. | |
1.8cm | |
25cm | |
Polystyrene | |
Small |
Polyethylene Copolymer (Blade), Polystyrene (Handle) | |
1.8 cm (Blade), 25 cm (Handle) | |
10 in. | |
Polyethylene Copolymer | |
Cell Scraper |
Promotions
-
Fisherbrand
Spend More, Get More on Fisherbrand Plastic Labware
Buy $500 or more in qualifying plastic labware and get up to $500 more at no additional cost.: 06/30/21
-
Fisherbrand
Buy $500, Get $500 in Plastic Cultureware
Purchase $500 in select Fisherbrand™ cultureware and get $500 more to spend on products at no additional cost.: 06/30/21