Please login to your online account to display your discounted pricing
Cartridge Kits for Milli-Q™ Pure Water Polishing Systems
High-quality filters fit Millipore™ Milli-Q three- or four-bowl systems—without expensive hardware or conversion kits
Manufacturer: thermo sci water  Y2753
 View more versions of this product
Catalog No. 09-025-20
  • / Each
Description & Specifications
Specifications
For Use With (Equipment) Milli-Q™ pure water polishing system
Material Polypropylene
Product Type Cartridge Kit
Includes Two Carbon, fout ion exchange cartridge, four hollow Fiber 0.2μm and two extra final filter