Fisherbrand™ Cartridge Kits for Milli-Q™ Pure Water Polishing Systems
High-quality filters fit Millipore™ Milli-Q three- or four-bowl systems—without expensive hardware or conversion kits
Supplier: Fisherbrand™ Y2752
Description
- Deliver bacteria-free Type I reagent-grade water for a variety of applications
- Cartridge housing: polypropylene
- Ultrasonically welded
- Ink-, solvent- and dye-free
- Choose from multicartridge kits with complete single or double change
- Double-change kits include two extra final filters for critical applications
Specifications
Polypropylene | |
Milli-Q™ pure water polishing system | |
One Carbon, two ion exchange, one organic extraction, one hollow Fiber 0.2μm and two extra final filters for critical applications | |
Cartridge Kit |