Please login to your online account to display your discounted pricing
Fisherbrand™ Cartridge Kits for Milli-Q™ Pure Water Polishing Systems
High-quality filters fit Millipore™ Milli-Q three- or four-bowl systems—without expensive hardware or conversion kits
Manufacturer: fisherbrand™  Y2751
 View more versions of this product
Catalog No. 09-025-22
  • / Each
Description & Specifications
Specifications
For Use With (Equipment) Milli-Q™ pure water polishing system
Product Type Cartridge Kit
Material Polypropylene
Includes One Prefilter, one Carbon, two ion exchange, one hollow Fiber 0.2μm and two extra final filters for critical applications