missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Burn-up Bin™ Biohazard Waste Boxes
Store flat, assemble in seconds.
Description
Store flat, assemble in seconds.
- Floor model box features integral handles for safety in use and transport
- Corrugated cardboard—single use
- 2 mil polyliner safely holds biohazard waste
- Tight-fitting lid has flap opening to accept material; to dispose, simply close lid and transfer filled box to incinerator
- Graphics and biohazard symbols readily identify contents, encourage proper use
- Replacement bags are not available for the Burn-Up Bin as boxes are to be incinerated after use
Specifications
Benchtop Biohazard Waste Box | |
Bench model | |
7 in. | |
8 x 8 x 10 in. (20 x 20 x 25 cm) | |
20 cm | |
Polyethylene | |
8 in. | |
2 mm |
Cardboard | |
17.8 cm | |
White | |
25 cm | |
20 cm | |
Quick identification for non-hazardous waste paper products | |
8 in. | |
10 in. |