Fisherbrand™ Brix Hydrometers
Used to measure the concentration of sugar solutions in refineries.
Manufacturer:  Fisherbrand™ 11608D
 View more versions of this product
Catalog No. 11-608D
Description
Description
  • Calibrated at 20°C
  • Permanently marked with individual serial numbers
For measuring the concentration of sugar solutions in refineries
Certifications
NIST™ traceable. A certificate is provided to indicate traceability to standards provided by NIST™.
Compliance
Accuracy meets or exceeds ASTM™ E 100 specifications, calibrated according to ASTM™ E 126 specifications.
Specifications
Specifications
0.1°
300 mm
29 to 41°
250 mL
NIST, ASTM E100, ASTM E126
300mL Borosilicate Glass cylinder (08-530J)
Measuring the concentration of Sugar solutions in refineries
SDS
Documents
Product Certifications
Accessories