Fisherbrand™ Brix Hydrometers
Used to measure the concentration of sugar solutions in refineries.
 View more versions of this product
Catalog No. 11-608B
Description
Description
  • Calibrated at 20°C
  • Permanently marked with individual serial numbers
For measuring the concentration of sugar solutions in refineries
Certifications
NIST™ traceable. A certificate is provided to indicate traceability to standards provided by NIST™.
Compliance
Accuracy meets or exceeds ASTM™ E 100 specifications, calibrated according to ASTM™ E 126 specifications.
Specifications
Specifications
0.1
300 mm
9 to 21°
Measuring the concentration of Sugar solutions in refineries
NIST, ASTM E100, ASTM E126
300mL Borosilicate Glass cylinder (08-530J)
Soda Lime Glass
250 mL
SDS
Documents
Documents
Accessories