Fisherbrand™ Borosilicate Glass Disposable Serological Pipets with Wide Tip, Standard Length
Sterile and plugged, multipack
Manufacturer:  Fisherbrand™ FS7210810110
 View more versions of this product
Catalog No. 13-678-35B
Description
Description
Sterile and plugged, multipack
Compliance
Calibrated “To Deliver/Blow Out,” Color-coded packaging on all pipets, except bulk packs, allows easy identification by size using ISO™ standard colors.
Specifications
Specifications
0.33 oz.
ASTM E714
Yellow
Yes
2mL
±0.4mL
10mL
Sterile
0.33 oz., 10mL
Orange
Yes
0.2mL
Wide
Serological Pipet
Borosilicate Glass
Documents
Accessories