Fisherbrand™ Borosilicate Glass Disposable Serological Pipets with Wide Tip, Standard Length
Sterile and plugged, multipack
Manufacturer:  Fisherbrand™ FS721085110
 View more versions of this product
Catalog No. 13-678-35A
Description
Description
Sterile and plugged, multipack
Chemically inert pipets have negative graduations as well as bidirectional scales.
  • Blue markings are easy to read
  • Packaging meets requirements of small-, medium-, or large-volume use
  • Individually wrapped pipets come in transparent plastic/paper wrappers for easy identification and tear-through, and are further overpacked in handy shelf container/dispensers; ideal for small volumes requiring sterility
  • Multipack with resealable flap for larger volume use permits easy access to several pipets at a time without contaminating unused pipets
  • Outer packaging is recyclable
Compliance
Calibrated “To Deliver/Blow Out,” Color-coded packaging on all pipets, except bulk packs, allows easy identification by size using ISO™ standard colors.
Specifications
Specifications
0.16 oz.
ASTM E714
Yellow
Yes
1mL
±0.2mL
5mL
Sterile
0.16 oz., 5mL
Blue
Yes
0.1mL
Wide
Serological Pipet
Borosilicate Glass
Documents
Accessories