missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ BloodBloc™ Biohazard Wipes, Liners and Rolls
Prevent leakage of fluids and chemicals to hands, gloves and work surfaces
Manufacturer: Fisherbrand™ F0667051
Description
Wipes and Liners are non-sterile and not intended for use on patients.
- Soft, white absorbent wipe with bright-orange polyethylene backing is impermeable to blood, body fluids and strong solvents including xylene, xylene substitutes, acetones, ketones, alcohols and formalin
- Nonwoven cellulose captures and retains fluid; nonlinting for use with delicate instrumentation such as blood counters
- Orange flexible impermeable plastic backing prevents soak-through of pathogens and chemicals to gloves which help minimize cross-contamination when lab personal open doors or touch surfaces
- Can also be used as mats and liners; rolls can be cut to desired length
- One time use only
Specifications
Heavy | |
20.32cm | |
Prevent soak through of blood, body fluids, tissue and chemicals to gloves and surfaces when handling tubes, specimens, plates. | |
22.86cm | |
Orange Biohazard wipes | |
Blood tubes, microbiology plates, pipettes, probes, specimen containers, instruments | |
8 in. |
8 in. | |
9 x 8 in. (22.86 x 20.32cm) | |
9 in. | |
Flexible bright orange Polyethylene backing | |
Yes | |
9 in. |