missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Blood Bank Crossmatch Holders

For typing, screening, and other blood bank applications.

Manufacturer:  Fisherbrand™ 1595654

 View more versions of this product

Catalog No. 14-810-5




Description

Description

  • Clear acrylic shelves with white bases and sides
  • Holds donor sample tubes as well as tubes for donor's serum and cell suspension, ABO, Rh, antibody grouping, and screening
  • Available in four sizes

24-tube capacity: Holds four 16mm O.D. sample tubes and twenty 10mm O.D. crossmatch tubes
29-tube capacity: Holds one 16mm sample tube, one 13mm pilot tube serum, and twenty-seven 10mm phenotype, genotype, and cross-match tubes
38-tube capacity: Holds thirty-eight tubes for five sets of tubes with printed identification on top panel

  • Set 1: One 16mm patient, one serum tube, and one cell suspension tube
  • Set 2: Four 10mm-14mm forwarding group drives
  • Set 3: Three reverse grouping tubes
  • Set 4: Sixteen major cross match tubes
  • Set 5: Twelve minor cross match tubes
90-tube capacity: Holds six 16mm O.D. donor sample tubes, eighty-four 10 to 13mm tubes for donor's serum and cell suspension, ABO, Rh, antibody grouping, and screening

Specifications

Specifications

Not Autoclavable
10 to 16 mm
Blood bank crossmatch tubes
6.35 cm
13.25 in.
Acrylic
5.25 in.
Tube Rack
Clear,Transparent,White
13.25 x 5.25 x 2.5 in. (33.6 x 13.3 x 6.35cm)
2.5 in.
10 to 16 mm Tubes
33.6 cm
90
13.3 cm
Documents