Fisherbrand™ Bitran™ Specimen Storage Bags
Choice of Saranex™ odor-control film for long-term storage or polyethylene for short-term storage
Supplier: Fisherbrand™ 4746S
Description
Choice of Saranex™ odor-control film for long-term storage or polyethylene for short-term storage
- Use for collecting, transporting and storing tissue specimens
- Minimize storage space and help to eliminate leakage, breakage and contamination
- Resealable double-lock zipper closure
- Designed for storage of tissues and fixatives and for use as mini-anaerobic environments
Specifications
Zip Seal | |
16 x 16 in. (40.6 x 40.6cm) | |
40.6 cm | |
40.6 cm |
Saranex Film | |
16 in. | |
16 in. |