Fisherbrand™ Bitran™ Specimen Storage Bags
Choice of Saranex™ odor-control film for long-term storage or polyethylene for short-term storage
Manufacturer:  Fisherbrand™ 4759PE
 View more versions of this product
Catalog No. 19-075-207
Description
Description
Choice of Saranex™ odor-control film for long-term storage or polyethylene for short-term storage
  • Use for collecting, transporting and storing tissue specimens
  • Minimize storage space and help to eliminate leakage, breakage and contamination
  • Resealable double-lock zipper closure
  • Designed for storage of tissues and fixatives and for use as mini-anaerobic environments
Specifications
Specifications
24 x 24 in. (61 x 61cm)
24 in.
61cm
61cm
24 in.
Zip closure
PE (Polyethylene)
Documents