Fisherbrand™ Biohazard Specimen Bags
Access pocket without opening sealed pouch
Manufacturer:  Fisherbrand™ FSB4011
 View more versions of this product
Catalog No. 19-075-388F
Description
Description
Access pocket without opening sealed pouch
  • Two-pouch bags constructed of polyethylene
  • 2mil-thick zipper-seal pouch holds specimen container
  • Separate 1.5mil-thick pocket for papers Each bag printed with biohazard graphics in orange and blue
Specifications
Specifications
10 x 10 in. (25 x 25cm)
FDA
White
Printed with biohazard graphics, zipper closure, 2mil thick pouch, 1.5mil thick pocket
25cm
0.279mm
25cm
2 mil
Zipper
Access pocket without opening sealed pouch
10 in.
PE (Polyethylene)
10 in.