Please login to your online account to display your discounted pricing

Fisherbrand™ Beta Blocks

Designed to hold 1.5mL microcentrifuge tubes

$249.51 - $254.53

Products
Catalog Number Mfr. No. Price Quantity    

12-009-12

 
fisherbrand™
172334
Each for $254.53

12-009-49

 
fisherbrand™
172344
Each for $249.51
Description & Specifications

  • Ideal for either benchtop use or long-term storage of Beta-emitting radioactive isotope samples
  • Made from solid acrylic block with 0.37 in. thick (1.0cm) acrylic cover