Please login to your online account to display your discounted pricing
Fisherbrand™ Basic Protection SMS Disposable Lab Coats — Cranberry
Durable splash and dust protection
$61.35 - $61.35
Specifications
Color Cranberry
View More Specs
Products
Catalog Number Mfr. No. Size Price Quantity    
17100859
fisherbrand™
17100859
Large Case of 30 Each for $61.35
Description & Specifications
Specifications
Color Cranberry
Single-use lab coats provide high durability and protection. Excellent particle retention and water vapor transmission (cool to wear).
  • Latex-free, three-layer SMS construction
  • Packed 30 lab coats/case
  • Available in a variety of colors
  • Knit cuffs hold sleeves in place; Standard collar
  • Three pockets and a five-snap closure
  • Fabric weight: 50g/m2
Note: Other materials, colors and options are available. See Fisherbrand Basic Protection Disposable Polypropylene Lab Coats with Elastic Cuffs, and Fisherbrand Basic Protection Disposable Lab Coats with Pockets. SMS coats are also available in White, Medical Blue, Teal, Purple, and Blueberry.