Fisherbrand™ Basic Protection Polypropylene Disposable Lab Coats with Elastic Cuffs
Latex-free spunbond polypropylene construction lab ware
$24.68 - $27.35
Specifications
Color White
Material Polypropylene
Closure Type Snap
Cuff Style Elastic
Collar Type Standard
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
17100801 Fisherbrand™
17100801
Small Case of 25 Each for $24.68
17100802 Fisherbrand™
17100802
Medium Case of 25 Each for $25.27
17100804 Fisherbrand™
17100804
X-Large Case of 25 Each for $26.77
17100806 Fisherbrand™
17100806
3X-Large Case of 25 Each for $27.35
17100807 Fisherbrand™
17100807
5X-Large Case of 25 Each for $27.35
17100805 Fisherbrand™
17100805
2X-Large Case of 25 Each for $27.35
17100803 Fisherbrand™
17100803
Large Case of 25 Each for $26.77
Specifications
Description
  • Single-use lab coat
  • Standard collar, no pockets
  • Packed 25 lab coats/case
  • Elastic cuffs hold sleeves in place
  • Five snaps for a secure closure
  • Fabric weight: 50g/m2
  • White
Note: Other materials, colors and options are available. See Fisherbrand Basic Protection Disposable Lab Coats with Pockets, and Fisherbrand Basic Protection Disposable SMS Lab Coats in a variety of colors.
Description & Specifications
Specifications
White
Snap
Standard
Polypropylene
Elastic
Yes