Please login to your online account to display your discounted pricing
Fisherbrand™ General Protection Disposable SMS White Lab Coats
Single-use lab coats provide high durability and splash/dust protection
$17.23 - $17.94
Specifications
Collar Type Standard
Cuff Style Knit
Closure Type Snap
Latex Free Yes
Color White
View More Specs
Products
Catalog Number Mfr. No. Length (English) Size Price Quantity    
17-100-809A
fisherbrand™
17100809A
35 in. Small Pack of 10 Each for $17.23
Case of 3 Pack for $51.69
17-100-810A
fisherbrand™
17100810A
38 in. Medium Pack of 10 Each for $17.61
Case of 3 Pack for $52.83
17-100-811A
fisherbrand™
17100811A
39 in. Large Pack of 10 Each for $17.61
Case of 3 Pack for $52.83
17-100-812A
fisherbrand™
17100812A
41 in. X-Large Pack of 10 Each for $17.61
Case of 3 Pack for $52.83
17-100-813A
fisherbrand™
17100813A
43 in. 2X-Large Pack of 10 Each for $17.94
Case of 3 Pack for $53.82
17-100-814A
fisherbrand™
17100814A
45 in. 3X-Large Pack of 10 Each for $17.94
Case of 3 Pack for $53.82
17-100-815A
fisherbrand™
17100815A
50 in. 5X-Large Pack of 10 Each for $17.94
Case of 3 Pack for $53.82
Description & Specifications
Specifications
Collar Type Standard
Cuff Style Knit
Closure Type Snap
Latex Free Yes
Color White
Material SMS (Spunbound Melt-Blown Synthetic)
Disposable Yes
Product Type General protection; Three pockets
Roomy pockets for securly storing items like pens or small instruments
Latex-free, three-layer SMS construction
Packed 10 per pack; 30 per case
Excellent particle retention and water vapor transmission
Available in a variety of colors
Knit cuffs hold sleeves in place; standard collar
Three pockets and a five-snap closure
Fabric weight: 50g/m
Note: Other materials, colors and options are available. See Fisherbrand Basic Protection Disposable Polypropylene Lab Coats with Elastic Cuffs, and Fisherbrand Basic Protection Disposable Lab Coats with Pockets. SMS coats are also available in Medical Blue, Teal, Purple, Cranberry and Blueberry.