Please login to your online account to display your discounted pricing
Fisherbrand™ General Protection Disposable SMS Teal Lab Coats
Single-use lab coats provide high durability and splash/dust protection
$17.61 - $18.34
Specifications
Collar Type Standard
Cuff Style Knit
Closure Type Snap
Latex Free Yes
Color Teal
View More Specs
Products
Catalog Number Mfr. No. Length (English) Size Price Quantity    
17-100-817A
fisherbrand™
17100817A
35 in. Small Pack of 10 Each for $17.61
Case of 3 Pack for $52.83
17-100-818A
fisherbrand™
17100818A
38 in. Medium Pack of 10 Each for $17.99
Case of 3 Pack for $53.97
17-100-819A
fisherbrand™
17100819A
39 in. Large Pack of 10 Each for $17.99
Case of 3 Pack for $53.97
17-100-820A
fisherbrand™
17100820A
41 in. X-Large Pack of 10 Each for $17.99
Case of 3 Pack for $53.97
17-100-821A
fisherbrand™
17100821A
43 in. 2X-Large Pack of 10 Each for $18.32
Case of 3 Pack for $54.96
17-100-822A
fisherbrand™
17100822A
45 in. 3X-Large Pack of 10 Each for $18.32
Case of 3 Pack for $54.96
17-100-823A
fisherbrand™
17100823A
50 in. 5X-Large Pack of 10 Each for $18.34
Case of 3 Pack for $55.02
Description & Specifications
Specifications
Collar Type Standard
Cuff Style Knit
Closure Type Snap
Latex Free Yes
Color Teal
Material SMS (Spunbound Melt-Blown Synthetic)
Disposable Yes
Product Type General protection; Three pockets
Roomy pockets for securly storing items like pens or small instruments
Latex-free, three-layer SMS construction
Packed 10 per pack; 30 per case
Excellent particle retention and water vapor transmission
Available in a variety of colors
Knit cuffs hold sleeves in place; standard collar
Three pockets and a five-snap closure
Fabric weight: 50g/m
Note: Other materials, colors and options are available. See Fisherbrand Basic Protection Disposable Polypropylene Lab Coats with Elastic Cuffs, and Fisherbrand Basic Protection Disposable Lab Coats with Pockets. SMS coats are also available in White, Medical Blue, Purple, Cranberry and Blueberry.