Please login to your online account to display your discounted pricing
Fisherbrand™ Basic Protection Disposable Polypropylene White Lab Coats
Single-use lab coats provide economical, lightweight and breathable protection
$14.15 - $14.61
Specifications
Disposable Yes
Material Polypropylene
Closure Type Snap
Collar Type Standard
Cuff Style Knit
View More Specs
Products
Catalog Number Mfr. No. Size Price Quantity    
17-100-881A
fisherbrand™
17100881A
Small Pack of 10 Each for $14.15
Case of 3 Pack for $42.45
17-100-882A
fisherbrand™
17100882A
Medium Pack of 10 Each for $14.39
Case of 3 Pack for $43.17
17-100-883A
fisherbrand™
17100883A
Large Pack of 10 Each for $14.39
Case of 3 Pack for $43.17
17-100-884A
fisherbrand™
17100884A
X-Large Pack of 10 Each for $14.39
Case of 3 Pack for $43.17
17-100-885A
fisherbrand™
17100885A
2X-Large Pack of 10 Each for $14.61
Case of 3 Pack for $43.83
17-100-886A
fisherbrand™
17100886A
3X-Large Pack of 10 Each for $14.61
Case of 3 Pack for $43.83
17-100-887A
fisherbrand™
17100887A
5X-Large Pack of 10 Each for $14.61
Case of 3 Pack for $43.83
Description & Specifications
Specifications
Disposable Yes
Material Polypropylene
Closure Type Snap
Collar Type Standard
Cuff Style Knit
No. of Pockets 3
Latex Free Yes
Color White
Roomy pockets for securely storing items like pens or small instruments
Latex-free spunbond polypropylene construction
High level of breathability
Also available in blue color (Cat. Nos. 17-100-889A to -895A)
Knit cuffs hold sleeves in place
Three pockets and a five-snap closure
Standard collar
Disposable
Fabric weight: 48g/m
10/Pk; 30/Cs.
Note: Other materials, colors and options are available. See Fisherbrand Basic Protection Disposable Polypropylene Lab Coats with Elastic Cuffs, and Fisherbrand Basic Protection Disposable SMS Lab Coats in a variety of colors.