Please login to your online account to display your discounted pricing
Fisherbrand™ Basic Protection Disposable Polypropylene Medical Blue Lab Coats
Single-use lab coats provide economical, lightweight and breathable protection
$13.85 - $14.30
Specifications
Disposable Yes
Material Polypropylene
Closure Type Snap
Collar Type Standard
Cuff Style Knit
View More Specs
Products
Catalog Number Mfr. No. Size Price Quantity    
17-100-889A
fisherbrand™
17100889A
Small Pack of 10 Each for $13.85
Case of 3 Pack for $41.55
17-100-890A
fisherbrand™
17100890A
Medium Pack of 10 Each for $14.08
Case of 3 Pack for $42.24
17-100-891A
fisherbrand™
17100891A
Large Pack of 10 Each for $14.08
Case of 3 Pack for $42.24
17-100-893A
fisherbrand™
17100893A
2X-Large Pack of 10 Each for $14.30
Case of 3 Pack for $42.90
17-100-892A
fisherbrand™
17100892A
X-Large Pack of 10 Each for $14.08
Case of 3 Pack for $42.24
Description & Specifications
Specifications
Disposable Yes
Material Polypropylene
Closure Type Snap
Collar Type Standard
Cuff Style Knit
No. of Pockets 3
Latex Free Yes
Color Blue
Roomy pockets for securly storing items like pens or small instruments
Latex-free spunbond polypropylene construction
High level of breathability
Also available in white color (Cat. Nos. 17-100-881A to -887A)
Knit cuffs hold sleeves in place
Three pockets and a five-snap closure
Standard collar
Disposable
Fabric weight: 48g/m
10/Pk; 30/Cs.
Note: Other materials, colors and options are available. See Fisherbrand Basic Protection Disposable Polypropylene Lab Coats with Elastic Cuffs, and Fisherbrand Basic Protection Disposable SMS Lab Coats in a variety of colors.